missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis[2,5-Dimethoxy-4-(p-nitrophenylazo)benzenediazonium] Chlorozincate, TRC
Specifications
Specifications
| Chemical Name or Material | Bis[2,5-Dimethoxy-4-(p-nitrophenylazo)benzenediazonium] Chlorozincate |
| CAS | 64071-86-9 |
| Molecular Formula | 2[C14 H12 N5 O4] . [Cl4 Zn] |
| Quantity | 10 g |
| Synonym | Fast Black K Salt Hemi(zinc chloride) Salt, Tetrachloro-bis[2,5-dimethoxy-4-[(4-nitrophenyl)azo]benzenediazonium] Zincate, 2,5-Dimethoxy-4-(4-nitrophenyl)diazenylbenzenediazonium Dichlorozinc, 2,5-Dimethoxy-4-[(4-nitrophenyl)azo]-tetrachlorozincate(2-) Benzenediazonium (2:1) |
| InChI Formula | InChI=1S/2C14H12N5O4.4ClH.Zn/c2*1-22-13-8-12(14(23-2)7-11(13)16-15)18-17-9-3-5-10(6-4-9)19(20)21;;;;;/h2*3-8H,1-2H3;4*1H;/q2*+1;;;;;+2/p-4 |
| SMILES | [Cl-][Zn+2]([Cl-])([Cl-])[Cl-].COc1cc([N+]#N)c(OC)cc1N=Nc2ccc(cc2)N(=O)=O.COc3cc([N+]#N)c(OC)cc3N=Nc4ccc(cc4)N(=O)=O |
| Molecular Weight (g/mol) | 835.77 |
| Formula Weight | 831.9824 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?