missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,4-dimethylphenyl) Phosphate, TRC

Product Code. 30333903
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30333903 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30333903 Supplier Toronto Research Chemicals Supplier No. TRCB415163250MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bis(2,4-dimethylphenyl) Phosphate
CAS 108437-78-1
Molecular Formula C16H19O4P
Quantity 250 mg
Synonym 2,4-Dimethyl-phenol Hydrogen Phosphate, 2,4-Dimethyl-phenol 1,1'-(Hydrogen Phosphate)
InChI Formula InChI=1S/C16H19O4P/c1-11-5-7-15(13(3)9-11)19-21(17,18)20-16-8-6-12(2)10-14(16)4/h5-10H,1-4H3,(H,17,18)
SMILES CC1=CC(C)=C(OP(OC2=C(C)C=C(C)C=C2)(O)=O)C=C1
IUPAC Name bis(2,4-dimethylphenyl) hydrogen phosphate
Molecular Weight (g/mol) 306.29
Formula Weight 306.1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.