missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,4,4-trimethylpentyl)phosphinic Acid, TRC

Product Code. 30135667
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135667 5 g 5g
30132816 50 g 50g
30007804 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135667 Supplier Toronto Research Chemicals Supplier No. TRCB5857085G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83411-71-6
Molecular Formula C16H35O2P
Molecular Weight (g/mol) 290.42
IUPAC Name bis(2,4,4-trimethylpentyl)phosphinic acid
SMILES CC(CC(C)(C)C)CP(=O)(O)CC(C)CC(C)(C)C

Specifications

Quantity 5 g
InChI Formula InChI=1S/C16H35O2P/c1-13(9-15(3,4)5)11-19(17,18)12-14(2)10-16(6,7)8/h13-14H,9-12H2,1-8H3,(H,17,18)
Formula Weight 290.2375
Chemical Name or Material Bis(2,4,4-trimethylpentyl)phosphinic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.