missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,3-dibromopropoxy)tetrabromobisphenol A, TRC

Product Code. 30341480
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30341480 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30341480 Supplier Toronto Research Chemicals Supplier No. TRCB426150100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetrabromobisphenol A dibromopropyl ether
CAS 21850-44-2
Molecular Formula C21 H20 Br8 O2
Quantity 100 mg
Synonym Tetrabromobisphenol A-bis(2,3-dibromopropyl ether), Tetrabromobisphenol A-dibromopropyl ether, Benzene, 1,1'-(1-methylethylidene)bis[3,5-dibromo-4-(2,3-dibromopropoxy)- (9CI, ACI), 1,1'-(1-Methylethylidene)bis[3,5-dibromo-4-(2,3-dibromopropoxy)benzene] (ACI), Propane, 2,2-bis[3,5-dibromo-4-(2,3-dibromopropoxy)phenyl]- (8CI), 1,1'-Isopropylidenebis[3,5-dibromo-4-(2,3-dibromopropoxy)benzene], 1,3-Dibromo-5-[2-[3,5-dibromo-4-(2,3-dibromopropoxy)phenyl]propan-2-yl]-2-(2,3-dibromopropoxy)benzene, 2,2-Bis[3,5-dibromo-4-(2,3-dibromopropoxy)phenyl]propane, 2,2-Bis[4-(2,3-dibromopropoxy)-3,5-dibromophenyl]propane, 2,2-Bis[4-(2,3-dibromopropyloxy)-3,5-dibromophenyl]propane, 2,2-Bis[[3,5-dibromo-4-(2,3-dibromopropyloxy)]phenyl]propane, 3,3',5,5'-Tetrabromobisphenol A bis(2,3-dibromopropyl) ether, 4,4'-Isopropylidenebis[2,6-dibromo-1-(2,3-dibromopropoxy)benzene], Bis(2,3-dibromopropoxy)tetrabromobisphenol A, Bromkal 66-8, D 5532, E-R 08N, FCP 680, FCP 680G, FG 3100, Fire Cut P 680, Fire Cut P 680G, Fire Guard 3100, Flame Cut 121K, Flame Cut 121R, FR 720, GX 5532, HP 800A, OBE, PE 68, PE 68 (fireproofing agent), Pyroguard SR 720, Pyroguard SR 720N, Saytex HP 800A, Saytex HP 800AG, SR 720, SR 720N, TBBPA-DBPE, Tetrabromobisphenol A 2,3-dibromopropyl ether, Tetrabromobisphenol A bis(2,3-dibromopropyl ether), TexFRon 9001, XZ 6800
InChI Formula InChI=1S/C21H20Br8O2/c1-21(2,11-3-15(26)19(16(27)4-11)30-9-13(24)7-22)12-5-17(28)20(18(29)6-12)31-10-14(25)8-23/h3-6,13-14H,7-10H2,1-2H3
SMILES CC(C)(c1cc(Br)c(OCC(Br)CBr)c(Br)c1)c2cc(Br)c(OCC(Br)CBr)c(Br)c2
IUPAC Name 1,3-dibromo-5-[2-[3,5-dibromo-4-(2,3-dibromopropoxy)phenyl]propan-2-yl]-2-(2,3-dibromopropoxy)benzene
Molecular Weight (g/mol) 943.61
Formula Weight 935.493
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.