missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,2,6,6-tetramethyl-4-piperidyl) sebacate, 98%

Product Code. 15440666
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15440666 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15440666 Supplier Thermo Scientific Alfa Aesar Supplier No. H54025.22
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 52829-07-9 | C28H52N2O4 | 480.73 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 52829-07-9
Melting Point 82°C to 85°C
Flash Point 216°C (421°F)
Molecular Formula C28H52N2O4
MDL Number MFCD00134709
Quantity 100 g
UN Number UN2811
Synonym bis 2,2,6,6-tetramethyl-4-piperidyl sebacate, bis 2,2,6,6-tetramethylpiperidin-4-yl decanedioate, tinuvin 770, sanol, sumisorb 577, tinuvin 770df, tinuvin 770ls, sanol 770, sanol ls 700, sanol ls 770
InChI Key RWMYPXKVMUFMKS-UHFFFAOYSA-N
SMILES CC1(C)CNCC(C)(C)C1OC(=O)CCCCCCCCC(=O)OC1C(C)(C)CNCC1(C)C
IUPAC Name bis(2,2,6,6-tetramethylpiperidin-4-yl) decanedioate
Molecular Weight (g/mol) 480.73
PubChem CID 164282
Formula Weight 480.72
Percent Purity 98%
Chemical Name or Material Bis(2,2,6,6-tetramethyl-4-piperidyl) sebacate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.