missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)copper(II), 99.9% (metals basis)

Product Code. 11399657
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11399657 1 g 1g
11309667 5 g 5g
11319667 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11399657 Supplier Thermo Scientific Alfa Aesar Supplier No. 041279.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 14040-05-2 | C22H38CuO4 | 430.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14040-05-2
Molecular Formula C22H38CuO4
Molecular Weight (g/mol) 430.09
MDL Number MFCD00058920
InChI Key JGMGCTZGOAONPQ-UHFFFAOYSA-N
Synonym copper bis 2,2,6,6-tetramethyl-3,5-heptanedionate, 2,2,6,6-tetramethyl-5-2,2,6,6-tetramethyl-5-oxohept-3-en-3-yl oxy cuprio oxy hept-4-en-3-one
PubChem CID 91663146
SMILES [Cu++].CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C

Specifications

Melting Point 198°C
Boiling Point 100°C (0.1 mmHg)(sublimation)
Quantity 1 g
IUPAC Name copper;5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one
Formula Weight 430.05
Percent Purity 99.9%
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)copper(II)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.