missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)calcium(II), 99.99% (metals basis), Thermo Scientific Chemicals

Catalog Number 11308718
Click to view available options
Quantity:
0.5 g
2 g
10 g
50 g
This item is not returnable. View return policy
118448-18-3
C22H38CaO4
406.62
MFCD00064757
DOOFPPIHJGRIGW-OLHSYEKISA-L
ca tmhd 2, calcium bis dipivaloylmethanate, bis 2,2,6,6-tetramethyl-3,5-heptanedionato calcium
14598209
calcium;(Z)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate;(E)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate
CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].[Ca+2]
This item is not returnable. View return policy
118448-18-3
C22H38CaO4
406.62
MFCD00064757
DOOFPPIHJGRIGW-OLHSYEKISA-L
ca tmhd 2, calcium bis dipivaloylmethanate, bis 2,2,6,6-tetramethyl-3,5-heptanedionato calcium
14598209
calcium;(Z)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate;(E)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate
CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].[Ca+2]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 202°C to 206°C
Boiling Point 205°C (0.1 mmHg)(sublimation)
Quantity 0.5 g
Sensitivity Hygroscopic
Formula Weight 406.57
Percent Purity 99.99%
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)calcium(II)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'