missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)barium(II) hydrate, 98+%, Thermo Scientific™

Product Code. 11362488
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11362488 1 g 1g
11352488 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11362488 Supplier Thermo Scientific Alfa Aesar Supplier No. 014539.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MOCVD of superconductors

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17594-47-7
Molecular Formula C22H38BaO4
Molecular Weight (g/mol) 503.87
MDL Number MFCD00058709
InChI Key VJRLFEOHKCDHBT-UHFFFAOYSA-N
Synonym barium-dpm, ba tmhd 2, bis 2,2,6,6-tetramethyl-3,5-heptanedionato barium ii, barium 2,2,6,6-tetramethyl-3,5-heptanedionate tetramer, barium bis 2,2,6,6-tetramethyl-3,5-heptanedionate hydrate, bis 2,2,6,6-tetramethyl-3,5-heptanedionate barium ii, barium-thd, barium bis dipivaloylmethanate, barium ii 2,2,6,6-tetramethylheptanedionate, bis 2,2,6,6-tetramethyl-3,5 heptanedionato barium ii
PubChem CID 17749137
IUPAC Name barium(2+);(Z)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate;hydrate
SMILES [Ba++].CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C

Specifications

Melting Point 172°C
Color White
Boiling Point 225°C (0.3 mmHg)
Quantity 1 g
UN Number UN1564
Sensitivity Hygroscopic
Formula Weight 503.85 (Anhydrous)
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)barium(II) hydrate
compliance-icons
Product Identifier
  • Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)barium(II)
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.