missing translation for 'onlineSavingsMsg'
Learn More

Bis(1H-inden-1-yl)dimethylsilane, 97%, Thermo Scientific™

Product Code. 10218713 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10218713

Brand: Thermo Scientific Acros 388480010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 18666-26-7
MDL Number MFCD00674015
InChI Key DXYTUIWIDDBVLU-UHFFFAOYSA-N
Synonym bis indenyl dimethylsilane, di 1h-inden-1-yl dimethylsilane, bis 1h-inden-1-yl-dimethylsilane, 1h-indene,1,1'-dimethylsilylene bis, bis 1h-inden-1-yl dimethylsilane, dimethylbisindenylsilane, acmc-1bvug, dimethyldi 1-indenyl silane, dimethylbis 1-indenyl silane, dimethylbis indenyl silane
PubChem CID 599429
IUPAC Name bis(1H-inden-1-yl)-dimethylsilane
SMILES C[Si](C)(C1C=CC2=CC=CC=C12)C3C=CC4=CC=CC=C34

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C20H20Si
Quantity 1 g
Molecular Weight (g/mol) 288.465
Formula Weight 288.46
Percent Purity ≥96.0%
Physical Form Crystalline Powder
Chemical Name or Material Bis(1H-inden-1-yl)dimethylsilane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.