missing translation for 'onlineSavingsMsg'
Learn More

Bis(1,5-cyclooctadiene)rhodium(I) hexafluoroantimonate

Product Code. 15436985
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15436985

Brand: Thermo Scientific Alfa Aesar 045523.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 130296-28-5 | C16H24F6RhSb | 555.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130296-28-5
Molecular Formula C16H24F6RhSb
Molecular Weight (g/mol) 555.02
MDL Number MFCD11113031
InChI Key BROZUPPJWNODCU-SXGOCSEPSA-H
Synonym bis 1,5-cyclooctadiene rhodium i hexafluoroantimonate
PubChem CID 16689296
SMILES [Rh+].F[Sb-](F)(F)(F)(F)F.C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1

Specifications

Color Brown
Quantity 1 g
UN Number UN3077
IUPAC Name bis(cycloocta-1,5-diene) hexafluorostibanuide rhodium
Formula Weight 555.01
Physical Form Crystalline Powder
Chemical Name or Material Bis (1,5-cyclooctadiene)rhodium (I) hexafluoroantimonate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.