missing translation for 'onlineSavingsMsg'
Learn More

Bis(1,5-cyclooctadiene)nickel(0), TRC

Product Code. 30336441
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30336441 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30336441 Supplier Toronto Research Chemicals Supplier No. TRCB423990250MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bis(1,5-cyclooctadiene)nickel(0)
CAS 1295-35-8
Molecular Formula C16 H20 Ni
Quantity 250 mg
Synonym 1,5-Cyclooctadiene Nickel Complex, Bis(1,5-cyclooctadiene)nickel, Bis(cyclooctadiene)nickel, Bis(cyclooctadiene)nickel(0), Bis(cyclooctadienyl)nickel, Bis(η4-1,5-cyclooctadiene)nickel, Bis-1,5-cyclooctadienylnickel, Bis[(1,2,5,6-η)-1,5-cyclooctadiene]nickel, Di(1,5-cyclooctadiene)nickel, Dicyclooctadiene Nickel, Nickel Bis(cyclooctadiene), Nickel Dicyclooctadiene
InChI Formula InChI=1S/2C8H10.Ni/c2*1-2-4-6-8-7-5-3-1;/h2*1,8H,2,4-5,7H2;
SMILES C1CC2=CCCC(=C1)[Ni]23C4=CCCC3=CCC4
IUPAC Name 9,9'-spirobi[9λ4-nickelabicyclo[3.3.1]nona-1,5-diene]
Molecular Weight (g/mol) 271.023
Formula Weight 270.092
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.