missing translation for 'onlineSavingsMsg'
Learn More

Bis(1,5-cyclooctadiene)nickel(0)

Product Code. 15421527
Change view
Click to view available options
Quantity:
0.5 g
2 g
10 g
Unit Size:
0.5g
10g
2g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15421527 10 g 10g
15401527 0.5 g 0.5g
15411527 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15421527 Supplier Thermo Scientific Alfa Aesar Supplier No. 039234.09

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1295-35-8 | C16H24Ni | 275.06 g/mol

Bis(1,5-cyclooctadiene)nickel(0) is used as a catalyst for the cycloaddition of 1,3-dienes and is used to catalyze the addition of allyl phenyl sulfide to alkynes leading to 1,4-dienes. Also, it is involved as a catalyst for asymmetric alpha-arylation and heteroarylation of ketones with chloroarenes, stereoselective borylative ketone-diene coupling, cycloaddition of benzamides with internal alkynes and methyl carboxylation of homopropargylic alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1295-35-8
Molecular Formula C16H24Ni
Molecular Weight (g/mol) 275.06
MDL Number MFCD00058902
InChI Key JRTIUDXYIUKIIE-KZUMESAESA-N
Synonym bis 1,5-cyclooctadiene nickel 0, bis 1,5-cyclooctadiene nickel, ni cod 2, ccris 5973, bis cyclooctadiene nickel 0, nickel, bis 1,5-cyclooctadiene, bis cyclooctadiene nickel, 1z,5z-cycloocta-1,5-diene; nickel, nickel biscod, pubchem16018
PubChem CID 6433264
SMILES [Ni].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1

Specifications

Melting Point 60°C (decomposition)
Color Yellow
Odor Odorless
Quantity 10 g
UN Number UN1325
Sensitivity Air and light sensitive
Solubility Information Soluble in benzene,toluene,terahydrofuran,ether,dimethyl formamide,hexamethylphosphoramide,N-methylpyrrolidinone. Insoluble in water.
IUPAC Name (1Z,5Z)-cycloocta-1,5-diene;nickel
Formula Weight 275.06
Physical Form Crystalline
Chemical Name or Material Bis(1,5-cyclooctadiene)nickel(0)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.