missing translation for 'onlineSavingsMsg'
Learn More

Bis(1,5-cyclooctadiene)nickel(0), 96%

Product Code. 11480680
Change view
Click to view available options
Quantity:
2 g
Unit Size:
2g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11480680 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11480680 Supplier Thermo Scientific Alfa Aesar Supplier No. 044157.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1295-35-8 | C16H24Ni | 275.06 g/mol

Catalyst for isomerization and hydrosilyltion of unsaturated compounds. Co-catalyst for oligomerization/cyclooligomerization or polymerization of alkenes, Bis(1,5-cyclooctadiene)nickel(0) is used as a catalyst for the cycloaddition of 1,3-dienes and used to catalyze the addition of allyl phenyl sulfide to alkynes leading to 1,4-dienes. It is involved as a catalyst for asymmetric alpha-arylation and heteroarylation of ketones with chloroarenes, stereoselective borylative ketone-diene coupling, cycloaddition of benzamides with internal alkynes and methyl carboxylation of homopropargylic alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Catalyst for isomerization and hydrosilyltion of unsaturated compounds
  • Co-catalyst for oligomerization/cyclooligomerization or polymerization of alkenes, butadiene
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1295-35-8
Molecular Formula C16H24Ni
Molecular Weight (g/mol) 275.06
MDL Number MFCD00058902
InChI Key JRTIUDXYIUKIIE-KZUMESAESA-N
Synonym bis 1,5-cyclooctadiene nickel 0, bis 1,5-cyclooctadiene nickel, ni cod 2, ccris 5973, bis cyclooctadiene nickel 0, nickel, bis 1,5-cyclooctadiene, bis cyclooctadiene nickel, 1z,5z-cycloocta-1,5-diene; nickel, nickel biscod, pubchem16018
PubChem CID 6433264
IUPAC Name (1Z,5Z)-cycloocta-1,5-diene;nickel
SMILES [Ni].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1

Specifications

Melting Point 60°C (decomposition)
Color Yellow
Odor Odorless
Quantity 2 g
UN Number UN1325
Sensitivity Air and light sensitive
Solubility Information Soluble in benzene,toluene,terahydrofuran,ether,dimethyl formamide,hexamethylphosphoramide,N-methylpyrrolidinone. Insoluble in water.
Formula Weight 275.06
Percent Purity 96%
Physical Form Powder
Chemical Name or Material Bis (1,5-cyclooctadiene)nickel (0)
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Bis(1,5-cyclooctadiene)nickel(0)
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
  • Flammable solid Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H228-Flammable solid.
  • H317-May cause an allergic skin reaction.
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
Supplemental information
  • MIXTURE LIST-Contain: Bis(1,5-cyclooctadiene)nickel

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.