missing translation for 'onlineSavingsMsg'
Learn More

Bis(1,3-dithian-2-yl)methane-d2, TRC

Product Code. 30342583
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30342583 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30342583 Supplier Toronto Research Chemicals Supplier No. TRCB43175210MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bis(1,3-dithian-2-yl)methane-d2
CAS 105479-87-6
Molecular Formula C9 2H2 H14 S4
Quantity 10 mg
InChI Formula InChI=1S/C9H16S4/c1-3-10-8(11-4-1)7-9-12-5-2-6-13-9/h8-9H,1-7H2/i7D2
SMILES [2H]C([2H])(C1SCCCS1)C2SCCCS2
IUPAC Name 2-[dideuterio(1,3-dithian-2-yl)methyl]-1,3-dithiane
Molecular Weight (g/mol) 254.5
Formula Weight 254.026
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.