missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Birch pollen Bet V1, Rabbit anti-Plant, Clone: BV16, Novus Biologicals™
Rabbit Monoclonal Antibody
£217.00 - £477.00
Specifications
| Antigen | Birch pollen Bet V1 |
|---|---|
| Clone | BV16 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot, ELISA |
| Applications | Western Blot, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18331960
|
Novus Biologicals
NBP2-81093-0.025MG |
0.025 mg |
£217.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18095124
|
Novus Biologicals
NBP2-81093-0.2MG |
0.2 mg |
£477.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Birch pollen Bet V1 Monoclonal specifically detects Birch pollen Bet V1 in Plant samples. It is validated for Western Blot, ELISA.Specifications
| Birch pollen Bet V1 | |
| 1 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Plant | |
| Allergen Bet v I-A, Allergen=Bet v 1-A, BETVI, BETVIA, Major pollen allergen Bet v 1-A | |
| IgG κ | |
| Protein A purified | |
| This antibody binds to Birch pollen Bet V1. |
| BV16 | |
| Western Blot, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS | |
| Birch pollen Bet V1-A, a major allergen of the European white birch. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title