missing translation for 'onlineSavingsMsg'
Learn More

Birch pollen Bet V1 Antibody (M0418) - Azide and BSA Free, R&D Systems™

Product Code. 18687107 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
Unit Size:
25µg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18687107 0.025 mg 25µg
18652169 0.05 mg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18687107 Supplier Novus Biologicals Supplier No. NBP3090120.025mg
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Human Monoclonal Antibody

Birch pollen Bet V1 Monoclonal specifically detects Birch pollen Bet V1 in Plant samples. It is validated for ELISA∣X-Ray Crystallography.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Birch pollen Bet V1
Applications ELISA, X-Ray Crystallography
Classification Monoclonal
Clone M0418
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS
Gene Alias Allergen Bet v I-A, Allergen=Bet v 1-A, BETVI, BETVIA, Major pollen allergen Bet v 1-A
Host Species Human
Immunogen Bet v 1-A, a major allergen of the European white birch.
Purification Method Affinity purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Plant
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgE λ
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.