Learn More
Birch pollen Bet V1 Antibody (M0418) - Azide and BSA Free, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals NBP3-09012-0.05mg
 View more versions of this product
Product Code. 18652169
  • £685.00 / 50µg
Description
Birch pollen Bet V1 Monoclonal antibody specifically detects Birch pollen Bet V1 in Plant samples. It is validated for ELISA, X-Ray Crystallography
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Birch pollen Bet V1
Monoclonal
1 mg/mL
PBS
Human
Affinity purified
RUO
Plant
Purified
ELISA, X-Ray Crystallography
M0418
Unconjugated
Allergen Bet v I-A, Allergen=Bet v 1-A, BETVI, BETVIA, Major pollen allergen Bet v 1-A
Bet v 1-A, a major allergen of the European white birch.
0.05 mg
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgE λ