missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Birch pollen Bet V1 Antibody (BV16), Alexa Fluor™ 405, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Birch pollen Bet V1 Monoclonal specifically detects Birch pollen Bet V1 in Plant samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
| Antigen | Birch pollen Bet V1 |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | BV16 |
| Conjugate | Alexa Fluor 405 |
| Dilution | Western Blot, ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Allergen Bet v I-A, Allergen=Bet v 1-A, BETVI, BETVIA, Major pollen allergen Bet v 1-A |
| Host Species | Rabbit |
| Immunogen | Birch pollen Bet V1-A, a major allergen of the European white birch. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?