missing translation for 'onlineSavingsMsg'
Learn More

BIRC8 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18414438 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18414438 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18414438 Supplier Novus Biologicals Supplier No. NBP176669JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

BIRC8 Polyclonal antibody specifically detects BIRC8 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen BIRC8
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias baculoviral IAP repeat containing 8, baculoviral IAP repeat-containing 8, baculoviral IAP repeat-containing protein 8, hILP2, IAP-like protein 2, ILP-2ILP2, Inhibitor of apoptosis-like protein 2, Testis-specific inhibitor of apoptosis
Host Species Rabbit
Immunogen Antibody was raised with against a synthetic peptide corresponding to amino acids near the amino terminus of human ILP-2. The immunogen is located within the first 50 amino acids of ILP-2. Amino Acid Squence: TGYEARLITFGT
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Tumor Suppressors, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 112401
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.