missing translation for 'onlineSavingsMsg'
Learn More

BIRC6 Antibody, DyLight 550, Novus Biologicals™

Product Code. 18413677 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18413677 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18413677 Supplier Novus Biologicals Supplier No. NBP281962R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

BIRC6 Polyclonal antibody specifically detects BIRC6 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen BIRC6
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias APOLLON, baculoviral IAP repeat containing 6, baculoviral IAP repeat-containing 6, baculoviral IAP repeat-containing protein 6, BRUCE, FLJ13786, KIAA1289FLJ13726, Ubiquitin-conjugating BIR domain enzyme apollon, ubiquitin-conjugating BIR-domain enzyme apollon
Host Species Rabbit
Immunogen BIRC6 antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human BIRC6. The immunogen is located within amino acids 4760 - 4810 of BIRC6. Amino Acid Squence: ADIQQYSSDKRVGR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 57448
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.