missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ BIRC4 HEK293 Cell Transient Overexpression Lysate (Nondenatured)

Product Code. 16100994
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy

Product Code. 16100994

Brand: Abnova™ L148T6.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human overexpression lysate

Specifications

Cell Line HEK293
For Use With (Application) Immunoprecipitation, Western Blot
Description BIRC4 HEK293 Cell Transient Overexpression Lysate(Non-Denatured)
Gene Accession No. BC032729
Lysis Buffer Modified RIPA Lysis Buffer:50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF. ;Modified RIPA Lysis Buffer:50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF.
Molecular Weight (g/mol) Theoretical M.W. 57kDa
Plasmid pCMV-BIRC4 full length
Quality Control Testing Transient overexpression cell lysate was tested with anti-BIRC4 antibody (Mfr. No. H00000331-M01) by Western Blot.
Quantity 100 μg
Storage Buffer In modified RIPA Lysis Buffer.
Host Species Human
Concentration 2 mg/mL
Content And Storage Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Show More Show Less
compliance-icons
Product Identifier
  • BIRC4 monoclonal antibody (M01), clone 2B3
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin sensitiser Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P233-Keep container tightly closed.
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P404-Store in a closed container.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : sodium phosphate dibasic, potassium phosphate monobasic
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.