missing translation for 'onlineSavingsMsg'
Learn More

Birabresib, MedChemExpress

Product Code. 16486788
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16486788 10 mg 10mg
16476906 100 mg 100mg
16459337 5 mg 5mg
16451196 200 mg 200mg
16422867 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16486788 Supplier Medchem Express Supplier No. HY15743
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Birabresib (OTX-015) is a potent bromodomain (BRD2/3/4) inhibitor with IC50s ranging from 92 to 112 nM.

Birabresib (OTX-015) is a potent bromodomain (BRD2/3/4) inhibitor with IC50s ranging from 92 to 112 nM.

Specifications

Chemical Name or Material Birabresib
CAS 202590-98-5
Color Light Brown
Purity Grade Notes Research
Molecular Formula C25H22ClN5O2S
Quantity 10 mg
Synonym OTX-015 MK-8628
Solubility Information DMSO : ≥ 49 mg/mL (99.60 mM)
SMILES O=C(NC1=CC=C(O)C=C1)C[C@H]2C3=NN=C(C)N3C4=C(C(C)=C(C)S4)C(C5=CC=C(Cl)C=C5)=N2
Molecular Weight (g/mol) 491.99
Formula Weight 491.99
Percent Purity 98.19%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.