missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biprofen, TRC
Specifications
Specifications
| Quantity | 1 g |
| Synonym | (2RS)-2-(Biphenyl-4-yl)propanoic Acid, Flurbiprofen Imp. A (EP), Flurbiprofen Related Compound A (USP), 2-(Biphenyl-4-yl)propionic acid, Biprofen, [1,1'-Biphenyl]-4-acetic acid, α-methyl-, 4-Biphenylacetic acid, α-methyl- (8CI), Hydratropic acid, p-phenyl- (6CI,7CI), α-Methyl[1,1'-biphenyl]-4-acetic acid, (±)-2-(4-Biphenyl)propionic acid, 16300Y1, 2-((1,1'-Biphenyl]-4-yl)propanoic acid, 2-(4-Biphenyl)propionic acid, 2-(4-Biphenylyl)propanoic acid, 2-(4-Biphenylyl)propionic acid, 2-(4-Phenylphenyl)propionic acid, NSC 16300 |
| InChI Formula | InChI=1S/C15H14O2/c1-11(15(16)17)12-7-9-14(10-8-12)13-5-3-2-4-6-13/h2-11H,1H3,(H,16,17) |
| Formula Weight | 226.0994 |
| Percent Purity | >95% |
| Chemical Name or Material | (2RS)-2-(biphenyl-4-yl)propanoic acid |
| Grade | HPLC |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?