missing translation for 'onlineSavingsMsg'
Learn More

Biphenylene, 99+%, Thermo Scientific Chemicals

Product Code. 10381474 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mg
250 mg
Packaging:
Glass bottle
Unit Size:
250mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10381474 50 mg Glass bottle 50mg
10712491 250 mg Glass bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10381474 Supplier Thermo Scientific Acros Supplier No. 279740500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 259-79-0
Molecular Formula C12H8
Molecular Weight (g/mol) 152.2
MDL Number MFCD00001110
InChI Key IFVTZJHWGZSXFD-UHFFFAOYSA-N
Synonym diphenylene, cyclobutadibenzene, dibenzocyclobutadiene, 1,1'-biphenylene, unii-0z64i7d5m2, 8ci 9ci, biphenylen, biphenylene, biphenylene, 99+%, pubchem9106
PubChem CID 9214
ChEBI CHEBI:33079
IUPAC Name biphenylene
SMILES C1=CC=C2C(=C1)C3=CC=CC=C23

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 109°C to 112°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 50 mg
Beilstein 05, 298
Solubility Information Solubility in water: 9.60mg/l (calc). Other solubilities: organic solvents general
Formula Weight 152.2
Percent Purity 99+%
Physical Form Needles
Chemical Name or Material Biphenylene, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • Biphenylene
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.