missing translation for 'onlineSavingsMsg'
Learn More

Biphenyl, 99%

Product Code. 11463983
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11463983 250 g 250g
11453983 1000 g 1000g
11473983 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11463983 Supplier Thermo Scientific Alfa Aesar Supplier No. A10265.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 92-52-4 | C12H10 | 154.21 g/mol

Biphenyl used in combination with Li metal for the generation of activated Ca metal. It is also used as important starting material for the production of polychlorinated biphenyls. It is also an intermediate for the production of a host of other organic compounds

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Biphenyl used in combination with Li metal for the generation of activated Ca metal. It is also used as important starting material for the production of polychlorinated biphenyls. It is also an intermediate for the production of a host of other organic compounds

Solubility
Soluble in alcohol and ether

Notes
Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents. keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-52-4
Molecular Formula C12H10
Molecular Weight (g/mol) 154.21
MDL Number MFCD00003054
InChI Key ZUOUZKKEUPVFJK-UHFFFAOYSA-N
Synonym biphenyl, phenylbenzene, diphenyl, 1,1'-diphenyl, bibenzene, lemonene, xenene, phenador-x, tetrosin ly, carolid al
PubChem CID 7095
ChEBI CHEBI:17097
IUPAC Name 1,1'-biphenyl
SMILES C1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 69°C to 72°C
Density 0.99
Boiling Point 254°C to 255°C
Flash Point 113°C (235°F)
Odor Pleasant
Quantity 250 g
UN Number UN3077
Beilstein 1634058
Merck Index 14,3314
Solubility Information Soluble in alcohol and ether
Formula Weight 154.21
Percent Purity 99%
Chemical Name or Material Biphenyl
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Biphenyl
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Biphenyl

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.