missing translation for 'onlineSavingsMsg'
Learn More

Biphenyl-4-carboxaldehyde, 98+%

Product Code. 11399419
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11399419 5 g 5g
11452528 25 g 25g
11462528 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11399419 Supplier Thermo Scientific Alfa Aesar Supplier No. A15839.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3218-36-8 | C13H10O | 182.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3218-36-8
Molecular Formula C13H10O
Molecular Weight (g/mol) 182.22
MDL Number MFCD00006947
InChI Key ISDBWOPVZKNQDW-UHFFFAOYSA-N
Synonym 4-biphenylcarboxaldehyde, 4-biphenylaldehyde, p-phenylbenzaldehyde, 1,1'-biphenyl-4-carbaldehyde, biphenyl-4-carboxaldehyde, 4-formylbiphenyl, p-biphenylylaldehyde, 1,1'-biphenyl-4-carboxaldehyde, p-biphenylaldehyde, p-biphenylcarboxaldehyde
PubChem CID 76689
IUPAC Name 4-phenylbenzaldehyde
SMILES O=CC1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 57°C to 61°C
Density 1.107
Boiling Point 184°C (11 mmHg)
Flash Point 172°C (341°F)
Quantity 5 g
Beilstein 606693
Sensitivity Air Sensitive
Formula Weight 182.22
Percent Purity ≥98%
Chemical Name or Material Biphenyl-4-carboxaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.