missing translation for 'onlineSavingsMsg'
Learn More

Biphenyl-2-carboxylic acid, 98%

Product Code. 11459922
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459922 5 g 5g
11469922 25 g 25g
11479922 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11459922 Supplier Thermo Scientific Alfa Aesar Supplier No. A12049.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 947-84-2 | C13H10O2 | 198.221 g/mol

It is used as a pharmaceutical intermediate. he reaction of 2?-substituted biphenyl-2-carboxylic acids (where the 2?-substituent is H, CO2H, NO2, Cl, OMe, or CO2Me) with lead tetra-acetate in refluxing benzene solution, under a nitrogen atmosphere, affords 3,4-benzocoumarin as a major organic product.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate. he reaction of 2′-substituted biphenyl-2-carboxylic acids (where the 2′-substituent is H, CO2H, NO2, Cl, OMe, or CO2Me) with lead tetra-acetate in refluxing benzene solution, under a nitrogen atmosphere, affords 3,4-benzocoumarin as a major organic product.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 947-84-2
Molecular Formula C13H10O2
Molecular Weight (g/mol) 198.221
MDL Number MFCD00002463
InChI Key ILYSAKHOYBPSPC-UHFFFAOYSA-N
Synonym 2-biphenylcarboxylic acid, 1,1'-biphenyl-2-carboxylic acid, o-phenylbenzoic acid, biphenyl-2-carboxylic acid, biphenylcarboxylic acid, 1,1'-biphenyl carboxylic acid, diphenyl-2-carboxylic acid, phenylbenzoic acid
PubChem CID 70357
IUPAC Name 2-phenylbenzoic acid
SMILES C1=CC=C(C=C1)C2=CC=CC=C2C(=O)O

Specifications

Melting Point 110°C to 114°C
Boiling Point 343°C to 344°C
Odor Odorless
Quantity 5 g
Beilstein 974075
Solubility Information Insoluble in water.
Formula Weight 198.22
Percent Purity 98%
Chemical Name or Material Biphenyl-2-carboxylic acid
compliance-icons
Product Identifier
  • Biphenyl-2-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: o-Phenylbenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.