missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biphenyl-2-carboxylic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
It is used as a pharmaceutical intermediate. he reaction of 2?-substituted biphenyl-2-carboxylic acids (where the 2?-substituent is H, CO2H, NO2, Cl, OMe, or CO2Me) with lead tetra-acetate in refluxing benzene solution, under a nitrogen atmosphere, affords 3,4-benzocoumarin as a major organic product.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a pharmaceutical intermediate. he reaction of 2′-substituted biphenyl-2-carboxylic acids (where the 2′-substituent is H, CO2H, NO2, Cl, OMe, or CO2Me) with lead tetra-acetate in refluxing benzene solution, under a nitrogen atmosphere, affords 3,4-benzocoumarin as a major organic product.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep away from oxidizing agents.
It is used as a pharmaceutical intermediate. he reaction of 2′-substituted biphenyl-2-carboxylic acids (where the 2′-substituent is H, CO2H, NO2, Cl, OMe, or CO2Me) with lead tetra-acetate in refluxing benzene solution, under a nitrogen atmosphere, affords 3,4-benzocoumarin as a major organic product.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep away from oxidizing agents.
Specifications
Melting Point | 110°C to 114°C |
Boiling Point | 343°C to 344°C |
Odor | Odorless |
Quantity | 100 g |
Beilstein | 974075 |
Solubility Information | Insoluble in water. |
Formula Weight | 198.22 |
Percent Purity | 98% |
Chemical Name or Material | Biphenyl-2-carboxylic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'