missing translation for 'onlineSavingsMsg'
Learn More

Biphenyl-2-boronic acid pinacol ester, 97%

Product Code. 15435037
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15435037 1 g 1g
15445037 5 g 5g
15455037 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15435037 Supplier Thermo Scientific Alfa Aesar Supplier No. H32320.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 914675-52-8 | C18H21BO2 | 280.174 g/mol

Biphenyl-2-boronic acid pinacol ester, is used as a main benefits of the use of boronic esters in Suzuki-Miyaura cross-couplings- Enabling the coupling of organoboronic acids that are sensitive to hydrolytic deboronation. It includes other additional application such as a number of chiral boronic esters, such as the ones illustrated hereafter, have been used as inducers in stereoselective reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Biphenyl-2-boronic acid pinacol ester, is used as a main benefits of the use of boronic esters in Suzuki-Miyaura cross-couplings- Enabling the coupling of organoboronic acids that are sensitive to hydrolytic deboronation. It includes other additional application such as a number of chiral boronic esters, such as the ones illustrated hereafter, have been used as inducers in stereoselective reactions.

Solubility
Insoluble in water. Soluble in organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 914675-52-8
Molecular Formula C18H21BO2
Molecular Weight (g/mol) 280.174
MDL Number MFCD14636459
InChI Key WCXWQEUBHZKNMQ-UHFFFAOYSA-N
Synonym biphenyl-2-boronic acid pinacol ester, 2-1,1'-biphenyl-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-2-biphenylyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 59207619
IUPAC Name 4,4,5,5-tetramethyl-2-(2-phenylphenyl)-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2C3=CC=CC=C3

Spécification

Melting Point 76°C to 79°C
Quantity 1 g
Solubility Information Insoluble in water. Soluble in organic solvents.
Formula Weight 280.17
Percent Purity 97%
Chemical Name or Material Biphenyl-2-boronic acid pinacol ester

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.