missing translation for 'onlineSavingsMsg'
Learn More

Biphenyl-2-boronic acid, 95%

Product Code. 15459069
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 15459069

Brand: Thermo Scientific Alfa Aesar L17547.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 4688-76-0 | C12H11BO2 | 198.028 g/mol

It is a substrate employed in a palladium-catalyzed cross-coupling with benzylic acetates providing diarylmethanes. It is a reactant involved in Suzuki-Miyaura cross-coupling reactions with arylhalides, dibromovinyl precursors, alkenyl tosylates and mesylates, and quinoline carboxylates. It is involved in intramolecular Friedel-Crafts alkylation for synthesis of chiral tetralins, hydroxylation to phenols and oxidative coupling with alkynes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a substrate employed in a palladium-catalyzed cross-coupling with benzylic acetates providing diarylmethanes. It is a reactant involved in Suzuki-Miyaura cross-coupling reactions with arylhalides, dibromovinyl precursors, alke
TRUSTED_SUSTAINABILITY

Specifications

CAS 4688-76-0
Melting Point 173°C to 194°C
Molecular Formula C12H11BO2
MDL Number MFCD00136929
Quantity 1 g
Beilstein 3031806
Synonym 2-biphenylboronic acid, biphenyl-2-boronic acid, biphenyl-2-ylboronic acid, 2-biphenylboronicacid, 2-phenylphenyl boranediol, 2-phenylphenyl boronic acid, 1,1'-biphenyl-2-ylboronic acid, 2-biphenylylboronic acid, 1,1'-biphenylboronic acid, 2-phenylbenzeneboronic acid
Solubility Information Insoluble in water.
InChI Key HYCYKHYFIWHGEX-UHFFFAOYSA-N
SMILES B(C1=CC=CC=C1C2=CC=CC=C2)(O)O
IUPAC Name (2-phenylphenyl)boronic acid
Molecular Weight (g/mol) 198.028
PubChem CID 4589187
Formula Weight 198.03
Percent Purity 95%
Chemical Name or Material Biphenyl-2-boronic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.