missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biperiden hydrochloride, MedChemExpress
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Description
Biperiden (KL 373) hydrochloride is a non-selective muscarinic receptor antagonist that competitively binds to M1 muscarinic receptors, thereby inhibiting acetylcholine and enhancing dopamine signaling in the central nervous system. Biperiden hydrochloride has the potential for the research of Parkinson's disease and other related psychiatric disorders.
Specifications
Specifications
| Chemical Name or Material | Biperiden hydrochloride |
| Physical Form | Solid |
| CAS | 1235-82-1 |
| Color | White |
| Quantity | 500 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C21H30ClNO |
| For Use With (Application) | Neuroscience-Neurodegeneration |
| Synonym | KL 373 hydrochloride |
| Solubility Information | DMSO : 20 mg/mL (57.48 mM; Need ultrasonic) ∣H2O : 5 mg/mL (14.37 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction