missing translation for 'onlineSavingsMsg'
Learn More

Bioymifi, MedChemExpress

Product Code. 16437518
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16437518

Brand: Medchem Express HY18377

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Bioymifi (DR5 Activator), a potent TRAIL receptor DR5 activator, binds to the extracellular domain (ECD) of DR5 with a Kd of 1.2 μM. Bioymifi can act as a single agent to induce DR5 clustering and aggregation, leading to apoptosis.

Bioymifi (DR5 Activator), a potent TRAIL receptor DR5 activator, binds to the extracellular domain (ECD) of DR5 with a Kd of 1.2 μM. Bioymifi can act as a single agent to induce DR5 clustering and aggregation, leading to apoptosis.

Specifications

Chemical Name or Material Bioymifi
CAS 1420071-30-2
Color Reddish Brown
Purity Grade Notes Research
Molecular Formula C22H12BrN3O4S
Quantity 10 mg
Synonym DR5 Activator
Solubility Information DMSO : 12.5 mg/mL (25.29 mM; Need ultrasonic)
SMILES O=C(/C(SC1=N)=C/C2=CC=C(O2)C3=CC(C(N4)=O)=C(C=C3)C4=O)N1C5=CC=C(C=C5)Br
Molecular Weight (g/mol) 494.32
Formula Weight 494.32
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.