missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bioymifi, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Description
Bioymifi (DR5 Activator), a potent TRAIL receptor DR5 activator, binds to the extracellular domain (ECD) of DR5 with a Kd of 1.2 μM. Bioymifi can act as a single agent to induce DR5 clustering and aggregation, leading to apoptosis.
Specifications
Specifications
| Chemical Name or Material | Bioymifi |
| CAS | 1420071-30-2 |
| Color | Reddish Brown |
| Purity Grade Notes | Research |
| Molecular Formula | C22H12BrN3O4S |
| Quantity | 100 mg |
| Synonym | DR5 Activator |
| Solubility Information | DMSO : 12.5 mg/mL (25.29 mM; Need ultrasonic) |
| SMILES | O=C(/C(SC1=N)=C/C2=CC=C(O2)C3=CC(C(N4)=O)=C(C=C3)C4=O)N1C5=CC=C(C=C5)Br |
| Molecular Weight (g/mol) | 494.32 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction