missing translation for 'onlineSavingsMsg'
Learn More

Biochrom Biowave 3+

Product Code. 17233064
Click to view available options
Includes:
Spec, Bluetooth Connectivity
Spec, Bluetooth, Printer
Spec, Built-in Serial Printer
Spectrophotometer
Interface:
Large Disp,Button UI,Bluetooth
Large Display, Button UI
Printer Included:
No
Yes
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17233064

Brand: Biochrom 80300739

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The Biowave 3+ offers all the capabilities of a research grade UV/Vis spectrophotometer. Expect exceptionally high resolution with the 3 nm bandwidth. The robust design and reasonable cost make it great for students.

  • UV-Vis Spectrophotometer with a 3 nm bandwidth
  • Features a USB port for data collection
  • Pre-programmed life science methodologies, including nucleic acid quantification (DNA, RNA and oligonucleotides), protein assays, and for cell culture density (OD600) measurements
  • Nucleic acid scans for purity checking
  • Low volume capability for use with microvolume cuvettes
  • Novel optics with a Xenon source for long lamp lifetime and low running costs
  • Choose to include a built-in serial printer, Bluetooth connectivity, or both
  • Lightweight and stackable for storage
  • Spectrophotometer has a 2 year warranty and the Xenon flash lamp has a 3 year warranty
  • IQ/OQ available on request

Specifications

Beam Center Height 15 mm
Depth (English) 15.3 in.
Depth (Metric) 390 mm
Detector Type Twin CMOS Array
Display Graphic LCD
Languages English, French, German, Spanish, Italian, Chinese, Japanese
Light Source Xenon Flash
Product Line Biowave 3+
Spectral Bandwidth 3 nm
Type UV-Vis Spectrophotometer
Voltage 100/115/220/230 V
Wavelength Range 190 to 1100 nm
For Use With (Application) UV/Vis Spectrophotometry
For Use With (Equipment) Microvolume Cuvettes
Height (English) 3.9 in.
Height (Metric) 100 mm
Includes Spec, Bluetooth Connectivity
Interface Large Disp,Button UI,Bluetooth
Monochromator Flat Grating
Noise ±0.005 peak to peak ±0.002 RMS
Photometric Linearity ±1.3% or ±0.008 A (Whichever is Greater at 546 nm)
Photometric Range -0.3 to 2.5 A, 0.3 to 199 %T
Photometric Repeatability ±0.002 A to 0.5 A (at 546 nm)
Printer Included No
Shape Round
Stray Light <0.5 %T 340 nm
Wavelength Accuracy ±2 nm
Wavelength Repeatability ±1 nm
Weight (English) 6.6 lb.
Weight (Metric) 3 kg
Width (English) 10 in.
Width (Metric) 260 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.