missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Biotinylated Recombinant Human VEGF 165 Protein

Product Code. 18778086
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
This item is not returnable. View return policy

Product Code. 18778086

Brand: R&D Systems™ BT10543025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Biotinylated via amines

When Recombinant Human VEGFR1/Flt-1 Fc Chimera Recombinant Human VEGF R1/Flt-1 Fc Chimera (Catalog # 321-FL ) is immobilized at 0.2 μg/mL (100 μL/well), Biotinylated Recombinant Human VEGF 165 (Catalog # BT10543) binds with an ED50 of 0.8-6.4 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_001165097.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 7422
Molecular Weight (g/mol) M.W.-observed: 22-27 kDa, under reducing conditions, M.W.-theoretical: 19 kDa
Quantity 25 μg
Source Spodoptera frugiperda, Sf 21 (baculovirus)-derived human VEGF protein Ala27-Arg191
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias MVCD1, VAS, vascular endothelial growth factor A, Vascular permeability factor, Vasculotropin, VEGFA, VEGF-A, VEGFMGC70609, VPF, VPFvascular endothelial growth factor
Conjugate Biotin
Reconstitution Reconstitute at 250 μg/mL in 4 mM HCl.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein VEGF
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.