missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Biotinylated Cynomolgus Fc gamma RIII / CD16 Protein, His, Avitag™ (BLI verified)

Product Code. 15866717
Change view
Click to view available options
Quantity:
200 μg
25 μg
Unit Size:
200µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15866717 25 μg 25µg
15856717 200 μg 200µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15866717 Supplier ACROBiosystems Supplier No. FC6C82E025UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Biotinylated BLI verified Protein

CD16 is a low affinity Fc receptor, and has been identified as Fc receptors FcγRIIIa (CD16a) and FcγRIIIb (CD16b). These receptors bind to the Fc portion of IgG antibodies. CD16 encoded by two different highly homologous genes in a cell type-specific manner.CD16 is found on the surface of natural killer cells, neutrophil polymorphonuclear leukocytes, monocytes and macrophages.CD16a antigen is also known as Low affinity immunoglobulin gamma Fc region receptor III-A, Fc-gamma RIII-alpha. CD16b is a low-affinity, GPI-linked receptor expressed by neutrophils and eosinophils, whereas CD16a is an intermediate affinity polypeptide-anchored transmembrane glycoprotein expressed natural killer cells, macrophages, subpopulation of T-cells, immature thymocytes and placentaltrophoblasts.CD16a is involved in phagocytosis, secretion of enzymes and inflammatory mediators, antibody­dependent cytotoxicity and clearance of immune complexes. Aberrant expression or mutations of CD16a is implicated in susceptibility to recurrent viral infections, systemic lupus erythematosus, and alloimmune neonatal neutropenia.

This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 25.7 kDa. The protein migrates as 36-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.

Specifications

Accession Number NP_001270121.1(R25K)
Molecular Weight (g/mol) 25.7 kDa
Quantity 25 μg
Source Biotinylated Cynomolgus CD16, His,Avitag (BLI verified) (FC6-C82E0) is expressed from human 293 cells (HEK293). It contains AA Gly 17 - Gln 208 (Accession # Q8SPW2-1).
Immunogen Gly 17 - Gln 208
Storage Requirements -20°C
Gene Alias FCGR3
Common Name Fc gamma RIII / CD16
Conjugate Precision Avi
Species Cynomolgus Monkey
Protein Tag His Tag and Avi Tag
Buffer PBS, pH7.4
Endotoxin Level Less than 1.0 EU / μg by the LAL method.
Expression System HEK293
Form Powder
Protein Subtype MABSol™ Biotin Labeled Proteins
Purity or Quality Grade >95% as determined by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.