missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Biotinylated Cynomolgus Fc gamma RIIB / CD32b Protein, His, Avitag™ (SPR & BLI verified)

Product Code. 16497349
Change view
Click to view available options
Quantity:
200 μg
25 μg
Unit Size:
200µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497349 200 μg 200µg
16407359 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16497349 Supplier ACROBiosystems Supplier No. CDBC82E4200ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Biotinylated Protein

Receptors for the Fc region of IgG (Fc γ R) are members of the Ig superfamily that function in the activation or inhibition of immune responses. Three classes of human Fc γ Rs: RI (CD64), RII (CD32), and RIII (CD16), which generate multiple isoforms, are recognized. There are three genes for human Fcγ RII /CD32 (A, B, and C) and one for mouse Fcγ RII B (CD32B). CD32 is a low affinity receptor for IgG. Low affinity immunoglobulin gamma Fc region receptor II-b (FCGR2B) is also known as CD32b, FCG2, IGFR2. CD32B is expressed on B cells and myeloid dendritic cells. Ligation of CD32B on B cells downregulates antibody production and may, in some circumstances, promote apoptosis. Co-ligation of CD32B on dendritic cells inhibits maturation and blocks cell activation. CD32B may also be a target for monoclonal antibody therapy for malignancies.

This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 23.6 kDa. The protein migrates as 30-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.

Specifications

Accession Number NP_001271060.1
Molecular Weight (g/mol) 23.6 kDa
Quantity 200 μg
Source Biotinylated Cynomolgus CD32b, His,Avitag (SPR and BLI verified) (CDB-C82E4) is expressed from human 293 cells (HEK293). It contains AA Ala 46 - Pro 224 (Accession # Q8SPW3-1).
Immunogen Ala 46 - Pro 224
Storage Requirements -20°C
Gene Alias FCGR2B,CD32b,CD32,FCG2,IGFR2,CDw32
Common Name Fc gamma RIIB / CD32b
Conjugate Precision Avi
Species Cynomolgus Monkey
Protein Tag His Tag and Avi Tag
Buffer PBS, pH7.4
Endotoxin Level Less than 1.0 EU / μg by the LAL method.
Expression System HEK293
Form Powder
Protein Subtype MABSol™ Biotin Labeled Proteins
Purity or Quality Grade >95% as determined by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.