Learn More
R&D Systems™ Biotinylated Alkyne
Description
Includes
- Highly specific to the azido group.
- Can be linked to the azido group through click chemistry.
- Versatile for detection through the biotin moiety.
Provided in 20mM Tris, pH 8.0
Specifications
Specifications
| Quantity | 500 μg |
| Molecular Formula | C19H31N3O5S |
| Formula Weight | 413.53 Da |