missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biotinidase/BTD Rabbit anti-Human, Clone: 6, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90315-50UL
This item is not returnable.
View return policy
Description
Biotinidase/BTD Monoclonal specifically detects Biotinidase/BTD in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Biotinidase/BTD | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 686 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 6 | |
| ELISA 1:5000-1:10000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:100-1:500 | |
| biotinase, biotinidase, EC 3.5.1, EC 3.5.1.12 | |
| Recombinant Human Biotinidase/BTD Protein | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction