missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biotinidase/BTD Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-14891
This item is not returnable.
View return policy
Description
Biotinidase/BTD Polyclonal specifically detects Biotinidase/BTD in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Biotinidase/BTD | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| biotinase, biotinidase, EC 3.5.1, EC 3.5.1.12 | |
| Rabbit | |
| 61 kDa | |
| 0.1 mL | |
| Signal Transduction | |
| 686 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| P43251 | |
| BTD | |
| Recombinant protein encompassing a sequence within the center region of human BTD. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction