missing translation for 'onlineSavingsMsg'
Learn More

Biotinidase/BTD Antibody (3B10-2B3), Novus Biologicals™

Product Code. 18397987 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18397987 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18397987 Supplier Novus Biologicals Supplier No. H00000686M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Biotinidase/BTD Monoclonal antibody specifically detects Biotinidase/BTD in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen Biotinidase/BTD
Applications Western Blot, ELISA, Immunoprecipitation
Classification Monoclonal
Clone 3B10-2B3
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunoprecipitation
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH12099
Gene Alias biotinase, biotinidase, EC 3.5.1, EC 3.5.1.12
Host Species Mouse
Immunogen BTD (AAH12099, 1 a.a. ~ 543 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MAHAHIQGGRRAKSRFVVCIMSGARSKLALFLCGCYVVALGAHTGEESVADHHEAEYYVAAVYEHPSILSLNPLALISRQEALELMNQNLDIYEQQVMTAAQKDVQIIVFPEDGIHGFNFTRTSIYPFLDFMPSPQVVRWNPCLEPHRFNDTEVLQRLSCMAIRGDMFLVANLGTKEPCHSSDPRCPKDGRYQFNTNVVFSNNGTLVDRYRKHNLYFEAAFDVPLKVDLITFDTPFAGRFGIFTCFDILFFDPAIRVLRDYKVKHVVYPTAWMNQLPLLAAIEIQKAFAVAFGINVLAANVHHPVLGMTGSGIHTPLESFWYHDMENPKSHLIIAQVAKNPVGLIGAENATGETDPSHSKFLKILSGDPYCEKDAQEVHCDEATKWNVNAPPTFHSEMMYDNFTLVPVWGKEGYLHVCSNGLCCYLLYERPTLSKELYALGVFDGLHTVHGTYYIQVCALVRCGGLGFDTCGQEITEATGIFEFHLWGNFSTSYIFPLFLTSGMTLEVPDQLGWENDHYFLRKSRLSSGLVTAALYGRLYERD
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 686
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.