missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biotin (Vitamin B7 or Vitamin H) Mouse anti-All species, Clone: BTN399, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Biotin (Vitamin B7 or Vitamin H) Monoclonal antibody specifically detects Biotin (Vitamin B7 or Vitamin H) in All species samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), In Situ Hybridization (ISH)
Specifications
Specifications
| Antigen | Biotin (Vitamin B7 or Vitamin H) |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), In Situ Hybridization (ISH) |
| Classification | Monoclonal |
| Clone | BTN399 |
| Concentration | 0.2 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.5-2 ug/ml, Flow Cytometry 0.5-1 ug/million cells, Immunofluorescence 0.5-1 ug/ml, Immunohistochemistry-Paraffin 0.5-1 ug/ml, In Situ Hybridization (ISH) |
| Formulation | 10 mM PBS with 0.05% BSA |
| Gene Alias | 58-85-5, coenzyme R, d-biotin, Vitamin B7, vitamin H |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?