missing translation for 'onlineSavingsMsg'
Learn More

Biotin (Vitamin B7 or Vitamin H) Antibody (BTN399), PerCP, Novus Biologicals™

Product Code. 18339641 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18339641 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18339641 Supplier Novus Biologicals Supplier No. NBP314084PCP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Biotin (Vitamin B7 or Vitamin H) Monoclonal antibody specifically detects Biotin (Vitamin B7 or Vitamin H) in All species samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Biotin (Vitamin B7 or Vitamin H)
Applications Flow Cytometry
Classification Monoclonal
Clone BTN399
Conjugate PerCP
Formulation PBS
Gene Alias 58-85-5, coenzyme R, d-biotin, Vitamin B7, vitamin H
Host Species Mouse
Immunogen Biotinylated sheep immunoglobulin
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species All species
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.