missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Biotin-Ubiquitin

Product Code. 11545016
Change view
Click to view available options
Quantity:
10 μg
100 μg
Unit Size:
100µg
10µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11545016 10 μg 10µg
11555016 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 11545016 Supplier Invitrogen™ Supplier No. PV4379

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Biotin-labeled ubiquitin for use as a general research tool, or in high throughput assays to monitor protein ubiquitination.

Biotin-labeled ubiquitin for use as a general research tool, or in high throughput assays to monitor protein ubiquitination. Due to the method in labeling the ubiquitin protein, all of the lysines with the protein are available for further conjugation to form poly-ubiquitin chains.

For use in combination with other LanthaScreen™ products including LanthaScreen™ Tb-Ubiquitin (Cat. No. PV4375), Fluorescein-Ubiquitin (PV4377), or a LanthaScreen™ antibody such as Tb-anti-biotin (PV4749).

TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 μg
Conjugate Biotin
Content And Storage Biotin labeled ubiquitin in HEPES buffered saline. Store at -20°C.
Detection Method Fluorescence
Excitation/Emission 340/490, 520 nm
For Use With (Equipment) Microplate Reader
Label or Dye Biotin
Label Type Biotin & Other Haptens
Readout End Point
Shipping Condition Dry Ice
Substrate Properties Protein-Based Substrate
Technique TR-FRET
Form Liquid
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.