missing translation for 'onlineSavingsMsg'
Learn More

(+)-Biotin (+)-Sulfoxide, TRC

Product Code. 30138066
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138066 1 g 1g
30009411 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138066 Supplier Toronto Research Chemicals Supplier No. TRCB3890501G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10406-89-0
Molecular Formula C10 H16 N2 O4 S
Molecular Weight (g/mol) 260.31
Synonym 1H-Thieno[3,4-d]imidazole-4-pentanoic acid, hexahydro-2-oxo-, 5-oxide, [3aS-(3aα,4β,5α,6aα)]-, Biotin, 5-oxide, (+)- (8CI), Biotin, D-S-oxide (6CI), (+)-Biotin (+)-sulfoxide, Biotin (+)-sulfoxide, Biotin d-sulfoxide, D-Biotin-d-sulfoxide
IUPAC Name 5-[(3aS,4S,6aR)-2,5-dioxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoic acid
SMILES OC(=O)CCCC[C@H]1[C@H]2NC(=O)N[C@H]2CS1=O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C10H16N2O4S/c13-8(14)4-2-1-3-7-9-6(5-17(7)16)11-10(15)12-9/h6-7,9H,1-5H2,(H,13,14)(H2,11,12,15)/t6-,7-,9-,17?/m0/s1
Formula Weight 260.0831
Chemical Name or Material (+)-Biotin sulfoxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.