missing translation for 'onlineSavingsMsg'
Learn More

(-)-Biotin Sulfoxide, TRC

Product Code. 30004500
Change view
Click to view available options
Quantity:
10 mg
50 mg
250 mg
Unit Size:
10mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004500 10 mg 10mg
30008506 50 mg 50mg
30007523 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004500 Supplier Toronto Research Chemicals Supplier No. TRCB38904510MG
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3376-83-8
Molecular Formula C10 H16 N2 O4 S
Molecular Weight (g/mol) 260.31
Synonym 1H-Thieno[3,4-d]imidazole-4-pentanoic acid, hexahydro-2-oxo-, 5-oxide, [3aS-(3aα,4β,5β,6aα)]-, Biotin, 5-oxide (7CI), Biotin, 5-oxide, (-)- (8CI), Biotin, L-S-oxide (6CI), (-)-Biotin sulfoxide, Biotin l-sulfoxide, Biotin sulfoxide
IUPAC Name 5-[(3aS,4R,6aR)-2,5-dioxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoic acid
SMILES OC(=O)CCCC[C@@H]1[C@H]2NC(=O)N[C@H]2CS1=O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C10H16N2O4S/c13-8(14)4-2-1-3-7-9-6(5-17(7)16)11-10(15)12-9/h6-7,9H,1-5H2,(H,13,14)(H2,11,12,15)/t6-,7+,9-,17?/m0/s1
Formula Weight 260.0831
Percent Purity >95%
Chemical Name or Material (-)-Biotin Sulfoxide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.