missing translation for 'onlineSavingsMsg'
Learn More

(+)-Biotin N-succinimidyl ester, 98%

Product Code. p-7059520
Click to view available options
Quantity:
50 mg
0.25 g
1 g
Unit Size:
0.25g
1g
50mg
This item is not returnable. View return policy

Product Code. 11430031

Brand: Thermo Scientific Alfa Aesar 044771.MF

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

In coupling reactions to amino acids, peptides and proteins through primary amines | CAS: 35013-72-0 | C14H19N3O5S | 341.382 g/mol

(+)-Biotin N-succinimidyl ester is used in coupling reactions to amino acids, peptides and proteins through primary amines. It is used as amino reactive biotin reagent and in the preparation of biotinylated surfaces or polypeptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35013-72-0
Molecular Formula C14H19N3O5S
Molecular Weight (g/mol) 341.382
MDL Number MFCD00078531
InChI Key YMXHPSHLTSZXKH-RVBZMBCESA-N
Synonym +-biotin n-hydroxysuccinimide ester, biotin-nhs, biotin nhs, 2,5-dioxopyrrolidin-1-yl 5-3as,4s,6ar-2-oxohexahydro-1h-thieno 3,4-d imidazol-4-yl pentanoate, n-hydroxysuccinimidobiotin, d-biotinyl-osu, biotin-osu, n-succinimidyl d-biotinate, bhse, biotin n-hydroxysuccinimide ester
PubChem CID 6710714
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoate
SMILES C1CC(=O)N(C1=O)OC(=O)CCCCC2C3C(CS2)NC(=O)N3

Specifications

Melting Point 212°C to 215°C
Color White
Quantity 50 mg
Beilstein 4720781
Solubility Information Soluble in Dimethylformamide ?50mg/mL (Stable for at least one month in dry DMF). Soluble in Dimethyl sulfoxide(DMSO).
Formula Weight 341.4
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (+)-Biotin N-succinimidyl ester

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.