missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MA5-11251 targets Biotin in Western blot, immunomicroscopy, in situ hybridization and flow cytometry applications and shows reactivity with All samples. The MA5-11251 immunogen is biotin conjugated to BSA.
Biotin Monoclonal antibody specifically detects Biotin in Chemical samples. It is validated for Flow Cytometry, Immunomicroscopy, In Situ Hybridization (ISH), Western Blot.
Specifications
Specifications
| Antigen | Biotin |
| Applications | Flow Cytometry, Immunomicroscopy, In Situ Hybridization (ISH), Western Blot |
| Classification | Monoclonal |
| Clone | BTN.4 |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 |
| Gene Alias | Vitamin B4 |
| Host Species | Mouse |
| Immunogen | Biotin conjugated to BSA |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?