missing translation for 'onlineSavingsMsg'
Learn More

Biotin, MedChemExpress

Product Code. 16460968
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16460968 5 g 5g
16422209 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16460968 Supplier Medchem Express Supplier No. HYB0511
 Restricted Item
Estimated Shipment: 05-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Biotin (Vitamin B7) is a water-soluble B vitamin and serves as a coenzyme for five carboxylases in humans, involved in the synthesis of fatty acids, isoleucine, and valine, and in gluconeogenesis. Biotin is necessary for cell growth, the production of fatty acids, and the metabolism of fats and amino acids.

Biotin (Vitamin B7) is a water-soluble B vitamin and serves as a coenzyme for five carboxylases in humans, involved in the synthesis of fatty acids, isoleucine, and valine, and in gluconeogenesis. Biotin is necessary for cell growth, the production of fatty acids, and the metabolism of fats and amino acids.

Specifications

Chemical Name or Material Biotin
CAS 58-85-5
Color White
Purity Grade Notes Research
Molecular Formula C10H16N2O3S
Quantity 5 g
Synonym Vitamin B7 Vitamin H D-Biotin
Solubility Information DMSO : 100 mg/mL (409.32 mM; Need ultrasonic) ∣H2O : 1 mg/mL (4.09 mM; Need ultrasonic)
SMILES O=C(O)CCCC[C@@H]1SC[C@]([C@]1([H])N2)([H])NC2=O
Molecular Weight (g/mol) 244.31
Formula Weight 244.31
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.