missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biotin-conjugated Human IgG Heavy chain Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
£119.00 - £295.00
Specifications
| Antigen | IgG Heavy chain |
|---|---|
| Clone | 1C6B4 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30235108
|
Proteintech
Biotin-67760-20UL |
20 μL |
£130.00 £119.00 / 20µL Save £11.00 8% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30236302
|
Proteintech
BIOTIN-67760-100UL |
100 μL |
£350.00 £295.00 / 100µL Save £55.00 15% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Human IgG Heavy chain antibody from Proteintech is a mouse monoclonal antibody generated with nature protein of Human IgG Heavy chain. This antibody recognizes human antigen. The Human IgG Heavy chain antibody has been validated for the following applications: WB analysis. This antibody is biotin.Specifications
| IgG Heavy chain | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| nature protein | |
| Primary | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 1C6B4 | |
| Western Blot | |
| Biotin | |
| Mouse | |
| Human | |
| IgG2a | |
| Protein A purification | |
| Conjugated Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title